Learn More
Raybiotech IncSupplier Diversity Partner Human SERPINA1 (Alpha-1-Antitrypsin) ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELHSERPINA11
For cell culture supernatants, plasma, and serum samples. Sensitivity: 2 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects human Serpin A1. Other species not determined.
Catalog No. 50-194-9711