Learn More
Novus Biologicals™ Human Serpin A6/Cortisol Binding Globulin ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP275053
Catalog No. NBP275053
Add to cart
Description
Description
The Human Serpin A6/Cortisol Binding Globulin ELISA Kit (Colorimetric) measures human Serpin A6/Cortisol Binding Globulin in serum, plasma and other biological fluids.
Specifications
Specifications
6.25 - 400 ng/mL
ELISA
ELISA Kit (Colorimetric)
Storage is content dependent.
Human
1 Kit
This kit recognizes Human CBG in samples. No significant cross-reactivity or interference between Human CBG and analogues was observed.
866
3.75 ng/mL
Serpin A6/Cortisol Binding Globulin
SERPINA6
Biological Fluids, Plasma, Serum