Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Human Serpin E1/PAI-1 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier: Novus Biologicals™ NBP183740
Description
The Human Serpin E1/PAI-1 ELISA Kit (Colorimetric) measures human Serpin E1/PAI-1 in cell culture supernatant, serum, plasma (edta, citrate, heparin).Specifications
78 - 5,000pg/mL | |
ELISA Kit | |
Human | |
5054 | |
Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human PAI-1, Biotin-Conjugate anti-human PAI-1 polyclonal antibody, Streptavidin-HRP, Human PAI-1 Standard lyophilized, 10000 pg/mL upon reconstitution, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films | |
Cancer | |
Endothelial plasminogen activator inhibitor, PAI1PAI-1, PAISerpin E1, PLANH1plasminogen activator inhibitor 1, serine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogenactivator inhibitor type 1), member 1, serpin peptidase inhibitor, clade E |
29.0pg/mL | |
Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum | |
ELISA | |
SERPINE1 | |
96 Tests | |
Storage of components varies. See protocol for specific instructions. | |
Serpin E1/PAI-1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction