Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Serum Albumin Chicken anti-Human, Polyclonal, Gallus

Chicken Polyclonal Antibody

Manufacturer:  Exalpha Biologicals IncSupplier Diversity Partner AHSA

Encompass

 View more versions of this product

Catalog No. 50-210-0633


Add to cart

Description

Description

Human Serum Albumin Polyclonal antibody specifically detects Human Serum Albumin in Human samples. It is validated for ELISA, Western Blot
Specifications

Specifications

Human Serum Albumin
Polyclonal
Unconjugated
ALB
Liquid
Chicken
IgY
0.5 mg
-20° C, Avoid Freeze/Thaw Cycles
Primary
213
ELISA, Western Blot
0.5-2 mg/mL
PBS with 0.075% sodium azide; pH
P02768
ALB; Alb1; Alb-1; albumin; albumin (32 AA); albumin (AA 34); albumin 1; Albza; ANALBA; cell growth inhibiting protein 42; DKFZp779N1935; FDAH; GIG20; GIG42; growth-inhibiting protein 20; PRO0883; PRO0903; PRO1341; PRO1708; PRO2044; PRO2619; PRO2675; Serum albumin; UNQ696/PRO1341
Human Serum Albumin
Affinity chromatography
RUO
Antibody
Polyclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit