Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Serum Albumin Chicken anti-Human, Polyclonal, Gallus Non-distribution product as customer accommodation.

Chicken Polyclonal Antibody

$176.64

Specifications

Antigen Human Serum Albumin
Concentration 0.5-2 mg/mL
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-210-0633 Non-distribution product as customer accommodation.
View Documents
Exalpha Biologicals IncSupplier Diversity Partner
AHSA
0.5 mg
Each for $176.64
Add to cart
 
50-210-0634 Non-distribution product as customer accommodation.
View Documents
Exalpha Biologicals IncSupplier Diversity Partner
AHSA10
10 mg
Each for $176.64
Add to cart
 
Description

Description

Human Serum Albumin Polyclonal antibody specifically detects Human Serum Albumin in Human samples. It is validated for ELISA, Western Blot
Specifications

Specifications

Human Serum Albumin
ELISA, Western Blot
Unconjugated
Chicken
Human
P02768
213
IgY
Primary
-20° C, Avoid Freeze/Thaw Cycles
ALB
0.5-2 mg/mL
Polyclonal
Liquid
RUO
PBS with 0.075% sodium azide; pH
ALB; Alb1; Alb-1; albumin; albumin (32 AA); albumin (AA 34); albumin 1; Albza; ANALBA; cell growth inhibiting protein 42; DKFZp779N1935; FDAH; GIG20; GIG42; growth-inhibiting protein 20; PRO0883; PRO0903; PRO1341; PRO1708; PRO2044; PRO2619; PRO2675; Serum albumin; UNQ696/PRO1341
Human Serum Albumin
Polyclonal
Affinity chromatography
Antibody
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit