Learn More
Raybiotech IncSupplier Diversity Partner Human sFRP-1 ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELHSFRP11
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.78 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human sFRP-1.
Catalog No. 50-194-9720