Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SH2B Partial ORF (AAH10704, 327 a.a. - 426 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00025970Q01S

 View more versions of this product

Catalog No. 89-961-767


Add to cart

Description

Description

This gene encodes a member of the SH2-domain containing mediators family. The encoded protein mediates activation of various kinases and may function in cytokine and growth factor receptor signaling and cellular transformation. Alternatively spliced transcript variants have been described. [provided by RefSeq]

Sequence: GKAKHLRLSLNEEGQCRVQHLWFQSIFDMLEHFRVHPIPLESGGSSDVVLVSYVPSSQRQQGREQAGSHAGVCEGDGCHPDASCTLMPFGASDCVTDHLP
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
GKAKHLRLSLNEEGQCRVQHLWFQSIFDMLEHFRVHPIPLESGGSSDVVLVSYVPSSQRQQGREQAGSHAGVCEGDGCHPDASCTLMPFGASDCVTDHLP
RUO
SH2B1
Wheat Germ (in vitro)
GST
AAH10704
Liquid
25970
SH2B (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp547G1110/FLJ30542/KIAA1299/SH2-B/SH2B
SH2B1
Yes
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit