Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SH3KBP1 Partial ORF (NP_114098.1, 224 a.a. - 308 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$460.50 - $709.50

Specifications

Name SH3KBP1 (Human) Recombinant Protein (Q01)
Recombinant Yes
For Use With (Application) Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
Molecular Weight 35.09kDa
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-961-821
View Documents
Abnova™
H00030011Q01L
25 ug
Each for $709.50
Add to cart
 
89-961-820
View Documents
Abnova™
H00030011Q01S
10 ug
Each for $460.50
Add to cart
 
Description

Description

CBL (MIM 165360) constitutively interacts with SH3 domain-containing proteins and, upon tyrosine phosphorylation, with SH2 domain-containing proteins. The SH3KBP1 gene encodes an 85-kD CBL-interacting protein that enhances tumor necrosis factor (MIM 191160)-mediated apoptotic cell death (Narita et al., 2001 [PubMed 11474197]).[supplied by OMIM]

Sequence: IKLRPRSIEVENDFLPVEKTIGKKLPATTATPDSSKTEMDSRTKSKDYCKVIFPYEAQNDDELTIKEGDIVTLINKDCIDVGWWE
Specifications

Specifications

SH3KBP1 (Human) Recombinant Protein (Q01)
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Yes
35.09kDa
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit