Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SIT1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF5016
This item is not returnable.
View return policy
Description
SIT1 Polyclonal specifically detects SIT1 in Human samples. It is validated for Western Blot.Specifications
SIT1 | |
Polyclonal | |
Unconjugated | |
Q9Y3P8 | |
SIT1 | |
E. coli-derived recombinant human SIT1 Gln65-Ser196 Accession # Q9Y3P8 | |
100 μg | |
Primary | |
Detects human SIT1 in direct ELISAs and Western blots. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
gp30/40, MGC125908, MGC125909, RP11-331F9.5, SHP2 interacting transmembrane adaptor, SHP2-interacting transmembrane adapter protein, SHP2-interacting transmembrane adaptor protein, signaling threshold regulating transmembrane adaptor 1, signaling threshold-regulating transmembrane adapter 1, SITMGC125910, suppression inducing transmembrane adaptor 1, Suppression-inducing transmembrane adapter 1 | |
Goat | |
Affinity Purified | |
RUO | |
27240 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction