Learn More
Novus Biologicals™ Human SLC6A3/DAT1 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier: Novus Biologicals™ NBP267271
Description
The Human SLC6A3/DAT1 ELISA Kit (Colorimetric) measures human SLC6A3/DAT1 in serum, plasma and other biological fluids.Specifications
0.16 - 10 ng/mL | |
This kit recognizes Human DAT in samples. No significant cross-reactivity or interference between Human DAT and analogues was observed. | |
Biological Fluids, Plasma, Serum | |
6531 | |
Human | |
Storage of components varies. See protocol for specific instructions. |
0.10 ng/mL | |
ELISA Kit (Colorimetric) | |
ELISA | |
SLC6A3 | |
1 Kit | |
SLC6A3/DAT1 |