Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SMPD1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF5348
This item is not returnable.
View return policy
Description
SMPD1 Polyclonal specifically detects SMPD1 in Human samples. It is validated for Western Blot, Simple Western.Specifications
SMPD1 | |
Polyclonal | |
Unconjugated | |
NP_000534 | |
SMPD1 | |
Chinese hamster ovary cell line CHO-derived recombinant human SMPD1 His62-Pro628 Accession # NP_000534 | |
100 μg | |
Primary | |
Detects human SMPD1 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant mouse SMPD1 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 50 ug/mL | |
Acid sphingomyelinase, ASMASE, ASMsphingomyelin phosphodiesterase, EC 3.1.4.12, NPD, sphingomyelin phosphodiesterase 1, acid lysosomal | |
Goat | |
Affinity Purified | |
RUO | |
6609 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction