Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SNCAIP Full-length ORF (AAH94759.1, 1 a.a. - 603 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$460.50 - $709.50

Specifications

Name SNCAIP (Human) Recombinant Protein (P01)
Recombinant Yes
Purification Method Glutathione Sepharose 4 Fast Flow
For Use With (Application) Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Molecular Weight 93kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-962-685
View Documents
Abnova™
H00009627P01L
25 ug
Each for $709.50
Add to cart
 
89-962-684
View Documents
Abnova™
H00009627P01S
10 ug
Each for $460.50
Add to cart
 
Description

Description

This gene encodes a protein containing several protein-protein interaction domains, including ankyrin-like repeats, a coiled-coil domain, and an ATP/GTP-binding motif. The encoded protein interacts with alpha-synuclein in neuronal tissue and may play a role in the formation of cytoplasmic inclusions and neurodegeneration. A mutation in this gene has been associated with Parkinson's disease. Alternatively spliced transcript variants encoding different isoforms of this gene have been described, but their full-length nature has yet to be determined. [provided by RefSeq]

Sequence: MTYLIQSHHSRRSQNCAEDVIRKTKTDQNGQLECVRWMVSETEAIAELSCSKDFPSLIHYAGCYGQEKILLWLLQFMQEQGISLDEVDQDGNSAVHVASQHGYLGCIQTLVEYGANVTMQNHAGEKPSQSAERQGHTLCSRYLVVVETCMSLASQVVKLTKQLKEQTVERVTLQNQLQQFLEAQKSEGKSLPSSPSSPSSPASRKSQWKSPDADDDSVAKSKPGVQEGIQVLGSLSASSRARPKAKDEDSDKILRQLLGKEISENVCTQEKLSLEFQDAQASSRNSKKIPLEKRELKLARLRQLMQRSLSESDTDSNNSEDPKTTPVRKADRPRPQPIVESVESMGSAESLHLMIKKHTLASGGRRFPFSIKASKSLDGHSPSPTSESSEPDLESQYPGSGSIPPNQPSGDPQQPSPDSTAAQKVATSPKSALKSPSSKRRTSQNLKLRVTFEEPVVQMEQPSLELNGEKDKDKGRTLQRTSTSNESGDQLKRPFGAFRSIMETLSGNQNNNNNYQAANQLKTSTLPLTSLGRKTDAKGNPASSASKGKNKAEMYSSCINLSSNMLIEEHLCNDTRHNDINRKMKKSYSIKHIAEPESKELFL
Specifications

Specifications

SNCAIP (Human) Recombinant Protein (P01)
Glutathione Sepharose 4 Fast Flow
93kDa
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit