Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SNF1LK2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF5737
This item is not returnable.
View return policy
Description
SNF1LK2/SIK2 Polyclonal specifically detects SNF1LK2/SIK2 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
SNF1LK2/SIK2 | |
Polyclonal | |
Unconjugated | |
Q9H0K1 | |
SIK2 | |
E. coli-derived recombinant human SNF1LK2 Ala663-Leu818 Accession # Q9H0K1 | |
100 μg | |
Primary | |
Detects human SNF1LK2 in Western blots. | |
Human | |
IgG |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
DKFZp434K1115, EC 2.7.11, EC 2.7.11.1, KIAA0781SIK-2, LOH11CR1I, QIK, QIKSNF1LK2, Qin-induced kinase, salt-inducible kinase 2, Salt-inducible protein kinase 2, salt-inducible serine/threonine kinase 2, serine/threonine-protein kinase SIK2, Serine/threonine-protein kinase SNF1-like kinase 2, SIK2, SNF1-like kinase 2 | |
Goat | |
Affinity Purified | |
RUO | |
23235 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction