Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human SNF8 Full-length ORF (NP_009172.2, 1 a.a. - 258 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00011267P01L
Description
SNF8, VPS25 (MIM 610907), and VPS36 (MIM 610903) form ESCRT-II (endosomal sorting complex required for transport II), a complex involved in endocytosis of ubiquitinated membrane proteins. SNF8, VPS25, and VPS36 are also associated in a multiprotein complex with RNA polymerase II elongation factor (ELL; MIM 600284) (Slagsvold et al., 2005 [PubMed 15755741]; Kamura et al., 2001 [PubMed 11278625]).[supplied by OMIM]
Sequence: MHRRGVGAGAIAKKKLAEAKYKERGTVLAEDQLAQMSKQLDMFKTNLEEFASKHKQEIRKNPEFRVQFQDMCATIGVDPLASGKGFWSEMLGVGDFYYELGVQIIEVCLALKHRNGGLITLEELHQQVLKGRGKFAQDVSQDDLIRAIKKLKALGTGFGIIPVGGTYLIQSVPAELNMDHTVVLQLAEKNGYVTVSEIKASLKWETERARQVLEHLLKEGLAWLDLQAPGEAHYWLPALFTDLYSQEITAEEAREALPSpecifications
NP_009172.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
55.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Dot3/EAP30/VPS22 | |
SNF8 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
11267 | |
SNF8 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MHRRGVGAGAIAKKKLAEAKYKERGTVLAEDQLAQMSKQLDMFKTNLEEFASKHKQEIRKNPEFRVQFQDMCATIGVDPLASGKGFWSEMLGVGDFYYELGVQIIEVCLALKHRNGGLITLEELHQQVLKGRGKFAQDVSQDDLIRAIKKLKALGTGFGIIPVGGTYLIQSVPAELNMDHTVVLQLAEKNGYVTVSEIKASLKWETERARQVLEHLLKEGLAWLDLQAPGEAHYWLPALFTDLYSQEITAEEAREALP | |
RUO | |
SNF8 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction