Learn More
Abnova™ Human SNX14 Partial ORF (AAH05110, 784 a.a. - 893 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | AAH05110 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 57231 |
Molecular Weight (g/mol) | 37.73kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-962-769
|
Abnova™
H00057231Q01L |
25 ug |
Each for $769.00
|
|
|||||
89-962-768
|
Abnova™
H00057231Q01S |
10 ug |
Each for $499.00
|
|
|||||
Description
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. The encoded protein also contains a regulator of G protein signaling (RGS) domain. Regulator of G protein signaling family members are regulatory molecules that act as GTPase activating proteins for G alpha subunits of heterotrimeric G proteins. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]
Sequence: QLFQEHRLVSLITLLRDAIFCENTEPRSLQDKQKGAKQTFEEMMNYIPDLLVKCIGEETKYESIRLLFDGLQQPVLNKQLTYVLLDIVIQELFPELNKVQKEVTSVTSWMSpecifications
AAH05110 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC13217/RGS-PX2 | |
SNX14 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
57231 | |
SNX14 (Human) Recombinant Protein (Q01) | |
QLFQEHRLVSLITLLRDAIFCENTEPRSLQDKQKGAKQTFEEMMNYIPDLLVKCIGEETKYESIRLLFDGLQQPVLNKQLTYVLLDIVIQELFPELNKVQKEVTSVTSWM | |
RUO | |
SNX14 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.