Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Human Sodium Iodide Symporter (hNIS) Ab-1, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Human Sodium Iodide Symporter (hNIS) Ab-1, Mouse Monoclonal Antibody.

$227.00 - $908.00

Specifications

Clone FP5A
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Host Species Murine
View More Specs

Products 7
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS1653P
View Documents
Thermo Scientific™
MS1653P
Purified Ab with BSA and Azide 1mL
Each for $861.50
Add to cart
 
MS1653P0
View Documents
Thermo Scientific™
MS1653P0
Purified Ab with BSA and Azide 100μL
Each for $227.00
Add to cart
 
MS1653P1
View Documents
Thermo Scientific™
MS1653P1
Purified Ab with BSA and Azide 500μL
Each for $525.50
Add to cart
 
MS1653P1ABX
View Documents
Thermo Scientific™
MS1653P1ABX
Ab without BSA/Azide (Frozen) 100μL
Each for $597.50
Add to cart
 
MS1653PABX
View Documents
Thermo Scientific™
MS1653PABX
Ab without BSA/Azide (Frozen) 200μL
Each for $908.00
Add to cart
 
MS1653R7
View Documents
Thermo Scientific™
MS1653R7
Ready-to-use for IHC (UVLP) 7mL
Each for $417.00
Add to cart
 
MS1653R1
View Documents
Thermo Scientific™
MS1653R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Provide accurate, reproducible results with the Thermo Scientific™ Human Sodium Iodide Symporter (hNIS) Ab-1, Mouse Monoclonal Antibody.

Human Sodium Iodide Symporter (hNIS) is responsible for iodide concentrating ability within thyroid follicular cells. It is a membrane bound glycoprotein with 13 membrane spanning domains and 14 extramembranous domains. It may represent an autoantigen in thyroid. Antibody to NIS showed strong immunostaining in Graves tissues, intermediate staining in papillary and follicular thyroid cancer, and no staining in Hurtle cell cancer or in nonthyroidal tissue.

Host Species: Mouse

Clone: FP5A

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with mouse and rat. Others not tested.

Epitope: aa 625-643

Immunogen: A synthetic peptide corresponding to aa 625-643 of hNIS protein

Molecular Weight: 97kDa

Positive Control: Thyroid

Cellular Localization: Membrane and cytoplasm

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Not verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

FP5A
Monoclonal
Murine
Primary
IgG2a kappa
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
RUO
A synthetic peptide corresponding to aa625-643 of hNIS protein
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit