Learn More
Abnova™ Human SPA17 Partial ORF (NP_059121, 51 a.a. - 149 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00053340Q01L
Description
This gene encodes a protein present at the cell surface. The N-terminus has sequence similarity to human cAMP-dependent protein kinase A (PKA) type II alpha regulatory subunit (RIIa) while the C-terminus has an IQ calmodulin-binding motif. The central portion of the protein has carbohydrate binding motifs and likely functions in cell-cell adhesion. The protein was initially characterized by its involvement in the binding of sperm to the zona pellucida of the oocyte. Recent studies indicate that it is also involved in additional cell-cell adhesion functions such as immune cell migration and metastasis. A retrotransposed pseudogene is present on chromosome 10q22
Sequence: EKTNFDPAEWGSKVEDRFYNNHAFEEQEPPEKSDPKQEESQISGKEEETSVTILDSSEEDKEKEEVAAVKIQAAFRGHIAREEAKKMKTNSLQNEEKEESpecifications
NP_059121 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
EKTNFDPAEWGSKVEDRFYNNHAFEEQEPPEKSDPKQEESQISGKEEETSVTILDSSEEDKEKEEVAAVKIQAAFRGHIAREEAKKMKTNSLQNEEKEE | |
RUO | |
SPA17 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
53340 | |
SPA17 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SP17/SP17-1 | |
SPA17 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.