Learn More
Abnova™ Human SPO11 Partial ORF (NP_036576, 291 a.a. - 395 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00023626Q01L
Description
Meiotic recombination and chromosome segregation require the formation of double-strand breaks (DSBs) in paired chromosome homologs. During meiosis in yeast, a meiotic recombination protein is covalently-linked to the 5' end of DSBs and is essential for the formation of DSBs. The protein encoded by this gene is similar in sequence and conserved features to the yeast meiotic recombination protein. The encoded protein belongs to the TOP6A protein family. Several transcript variants encoding different isoforms have been found for this gene, but the full-length nature of only two of them have been described. [provided by RefSeq]
Sequence: YGSMSMSFEAHHLTVPAIRWLGLLPSDLKRLNVPKDSLIPLTKRDQMKLDSILRRPYVTCQPFWRKEMEIMADSKMKAEIQALTFLSSDYLSRVYLPNKLKFGGWSpecifications
NP_036576 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.29kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
YGSMSMSFEAHHLTVPAIRWLGLLPSDLKRLNVPKDSLIPLTKRDQMKLDSILRRPYVTCQPFWRKEMEIMADSKMKAEIQALTFLSSDYLSRVYLPNKLKFGGW | |
RUO | |
SPO11 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
23626 | |
SPO11 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC39953 | |
SPO11 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.