Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human SSH3 Full-length ORF (AAH07709.1, 1 a.a. - 659 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00054961P01L
Description
The ADF (actin-depolymerizing factor)/cofilin family (see MIM 601442) is composed of stimulus-responsive mediators of actin dynamics. ADF/cofilin proteins are inactivated by kinases such as LIM domain kinase-1 (LIMK1; MIM 601329). The SSH family appears to play a role in actin dynamics by reactivating ADF/cofilin proteins in vivo (Niwa et al., 2002 [PubMed 11832213]).[supplied by OMIM]
Sequence: MALVTVSRSPPGSGASTPVGPWDQAVQRRSRLQRRQSFAVLRGAVLGLQDGGDNDDAAEASSEPTEKAPSEEELHGDQTDFGQGSQSPQKQEEQRQHLHLMVQLLRPQDDIRLAAQLEAPRPPRLRYLLVVSTREGEGLSQDETVLLGVDFPDSSSPSCTLGLVLPLWSDTQVYLDGDGGFSVTSGGQSRIFKPISIQTMWATLQVLHQACEAALGSGLVPGGSALTWASHYQERLNSEQSCLNEWTAMADLESLRPPSAEPGGSSEQEQMEQAIRAELWKVLDVSDLESVTSKEIRQALELRLGLPLQQYRDFIDNQMLLLVAQRDRASRIFPHLYLGSEWNAANLEELQRNRVTHILNMAREIDNFYPERFTYHNVRLWDEESAQLLPHWKETHRFIEAARAQGTHVLVHCKMGVSRSAATVLAYAMKQYECSLEQALRHVQELRPIARPNPGFLRQLQIYQGILTASRQSHVWEQKVGGVSPEEHPAPEVSTPFPPLPPEPEGGGEEKVVGMEESQAAPKEEPGPRPRINLRGVMRSISLLEPSLELESTSETSDMPEVFSSHESSHEEPLQPFPQLARTKGGQQVDRGPQPALKSRQSVVTLQGSAVVANRTQAFQEQEQGQGQGQGEPCISSTPRFRKVVRQASVHDSGEEGEASpecifications
AAH07709.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
99.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ10928/FLJ20515/SSH-3 | |
SSH3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
54961 | |
SSH3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MALVTVSRSPPGSGASTPVGPWDQAVQRRSRLQRRQSFAVLRGAVLGLQDGGDNDDAAEASSEPTEKAPSEEELHGDQTDFGQGSQSPQKQEEQRQHLHLMVQLLRPQDDIRLAAQLEAPRPPRLRYLLVVSTREGEGLSQDETVLLGVDFPDSSSPSCTLGLVLPLWSDTQVYLDGDGGFSVTSGGQSRIFKPISIQTMWATLQVLHQACEAALGSGLVPGGSALTWASHYQERLNSEQSCLNEWTAMADLESLRPPSAEPGGSSEQEQMEQAIRAELWKVLDVSDLESVTSKEIRQALELRLGLPLQQYRDFIDNQMLLLVAQRDRASRIFPHLYLGSEWNAANLEELQRNRVTHILNMAREIDNFYPERFTYHNVRLWDEESAQLLPHWKETHRFIEAARAQGTHVLVHCKMGVSRSAATVLAYAMKQYECSLEQALRHVQELRPIARPNPGFLRQLQIYQGILTASRQSHVWEQKVGGVSPEEHPAPEVSTPFPPLPPEPEGGGEEKVVGMEESQAAPKEEPGPRPRINLRGVMRSISLLEPSLELESTSETSDMPEVFSSHESSHEEPLQPFPQLARTKGGQQVDRGPQPALKSRQSVVTLQGSAVVANRTQAFQEQEQGQGQGQGEPCISSTPRFRKVVRQASVHDSGEEGEA | |
RUO | |
SSH3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction