Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ST7/LRP12 Antibody, R&D Systems™

Sheep Polyclonal Antibody
$200.50 - $549.50
Specifications
Antigen | ST7/LRP12 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
ST7/LRP12 Polyclonal specifically detects ST7/LRP12 in Human samples. It is validated for Western Blot.Specifications
ST7/LRP12 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Sheep | |
Human | |
DKFZp762O2113, FAM4A1, LRP12, Protein FAM4A1, Protein HELG, RAY1, SEN4, subfamily A, member 1, suppression of tumorigenicity 7, suppression of tumorigenicity 7 (breast), suppressor of tumorigenicity 7 protein, TSG7 | |
LRP12 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q9Y561 | |
29967 | |
Mouse myeloma cell line NS0-derived recombinant human ST7/LRP12 Asn28-Ile488 Accession # Q9Y561 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human ST7/LRP12 in direct ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant mouse ST7/LRP12 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title