Learn More
Abnova™ Human STAM2 Partial ORF (NP_005834, 416 a.a. - 525 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00010254Q01L
Description
The protein encoded by this gene is closely related to STAM, an adaptor protein involved in the downstream signaling of cytokine receptors, both of which contain a SH3 domain and the immunoreceptor tyrosine-based activation motif (ITAM). Similar to STAM, this protein acts downstream of JAK kinases, and is phosphorylated in response to cytokine stimulation. This protein and STAM thus are thought to exhibit compensatory effects on the signaling pathway downstream of JAK kinases upon cytokine stimulation. [provided by RefSeq]
Sequence: QSYSLGPDQIGPLRSLPPNVNSSVTAQPAQTSYLSTGQDTVSNPTYMNQNSNLQSATGTTAYTQQMGMSVDMSSYQNTTSNLPQLAGFPVTVPAHPVAQQHTNYHQQPLLSpecifications
NP_005834 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
QSYSLGPDQIGPLRSLPPNVNSSVTAQPAQTSYLSTGQDTVSNPTYMNQNSNLQSATGTTAYTQQMGMSVDMSSYQNTTSNLPQLAGFPVTVPAHPVAQQHTNYHQQPLL | |
RUO | |
STAM2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
10254 | |
STAM2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp564C047/Hbp/STAM2A/STAM2B | |
STAM2 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.