Learn More
Pepro Tech Inc Human sTRAIL/Apo2L ABTS ELISA Development Kit, 1Kit DFS Item
Manufacturer:  Pepro Tech Inc 900M141
This Human sTRAIL/Apo2L ELISA Kit includes: Capture Antibody, Detection Antibody, Protein Standard, and HRP-Conjugate. The components supplied in this kit are sufficient to assay human sTRAIL/Apo2L in approximately 200 ELISA plate wells.
Catalog No. 50-195-2615