Learn More
Human TAAR1, Synthetic Peptide, Invitrogen™
Synthetic Peptide
Manufacturer:  Invitrogen GST00001P
Catalog No. PIGST00001P
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Human TAAR1 Synthetic Peptide, GST00001P, from Invitrogen.
GST00001P is the immunizing peptide for the rabbit antibody to human trace amine-associated receptor 1 (OSR00079W).GST00001P may be used as a control in various immunology applications.After reconstitution, keep at -20ºC.Reconstitute with PBS or distilled water.
Specifications
Specifications
134864
Synthetic Peptide
Human
Control
peptide with no preservative
RUO
No preservative
Human TAAR1
Q96RJ0
Peptide
Human
purified
50μg
Lyophilized
Y
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Trace amine receptor 1, TaR-1, TA1, TAR1, TRAR1
TAAR1
Control