Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAAR2 Synthetic Peptide
Synthetic Peptide
Supplier: Thermo Scientific GST00002P
Description
Human TAAR2 Synthetic Peptide, GST00002P, from Invitrogen.
GST00002P is the immunizing peptide for the rabbit antibody to human trace amine-associated receptor 2 (OSR00080W).GST00002P may be used as a control in various immunology applications.Reconstitute with PBS or distilled water.Specifications
9287 | |
purified | |
Human TAAR2 | |
Synthetic Peptide | |
peptide with no preservative | |
Trace amine-associated receptor 2, TAAR2, GPR58 | |
Yes | |
Peptide | |
RUO - research use only |
Human | |
Q9P1P5 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized | |
Control | |
TAAR2 | |
No preservative | |
50 μg |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title