Learn More
Abnova™ Human TAS2R10 Partial ORF (AAH63585, 1 a.a. - 124 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00050839Q01S
Description
This gene product belongs to the family of candidate taste receptors that are members of the G-protein-coupled receptor superfamily. These proteins are specifically expressed in the taste receptor cells of the tongue and palate epithelia. They are organized in the genome in clusters and are genetically linked to loci that influence bitter perception in mice and humans. In functional expression studies, they respond to bitter tastants. This gene maps to the taste receptor gene cluster on chromosome 12p13. [provided by RefSeq]
Sequence: MLRVVEGIFIFVVVSESVFGVLGNGFIGLVNCIDCAKNKLSTIGFILTGLAISRIFLIWIIITDGFIQIFSPNIYASGNLIEYISYFWVIGNQSSMWFATSLSIFYFLKIANFSNYIFLWLKSGSpecifications
AAH63585 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
39.27kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MLRVVEGIFIFVVVSESVFGVLGNGFIGLVNCIDCAKNKLSTIGFILTGLAISRIFLIWIIITDGFIQIFSPNIYASGNLIEYISYFWVIGNQSSMWFATSLSIFYFLKIANFSNYIFLWLKSG | |
RUO | |
TAS2R10 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
50839 | |
TAS2R10 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC126811/MGC126813/T2R10/TRB2 | |
TAS2R10 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.