Learn More
Novus Biologicals™ Human Tau ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier: Novus Biologicals™ NBP262749
Description
The Human Tau ELISA Kit (Colorimetric) measures human Tau in serum, plasma and other biological fluids.Specifications
7.813--500 pg/mL | |
This kit recognizes Human MAP-tau in samples. No significant cross-reactivity or interference between Human MAP-tau and analogues was observed. | |
Biological Fluids, Plasma, Serum | |
4137 | |
Human | |
Storage of components varies. See protocol for specific instructions. |
4.688 pg/mL | |
ELISA Kit (Colorimetric) | |
ELISA | |
MAPT | |
1 Kit | |
Tau |