Learn More
Novus Biologicals™ Human Tau [p Ser739] ELISA Kit (Colorimetric)
SDP
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier:  Novus Biologicals™ NBP266711
Catalog No. NBP266711
Add to Cart
Description
Description
The Human Tau [p Ser739] ELISA Kit (Colorimetric) measures human Tau in serum, plasma and other biological fluids.
Specifications
Specifications
31.25 - 2000 pg/mL
This kit recognizes Human pMAPT/pTAU in samples. No significant cross-reactivity or interference between Human pMAPT/pTAU and analogues was observed.
Biological Fluids, Plasma, Serum
4137
Human
Storage of components varies. See protocol for specific instructions.
18.75 pg/mL
ELISA Kit (Colorimetric)
ELISA
MAPT
1 Kit
Tau
Recommendations
Recommendations