Learn More
Novus Biologicals™ Human Tenascin R ELISA Kit (Colorimetric)
Human Tenascin R ELISA Kit (Colorimetric) from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Manufacturer:  Novus Biologicals™ NBP276608
Catalog No. NBP276608
Add to cart
Description
Description
The Human Tenascin R ELISA Kit (Colorimetric) measures human Tenascin R in serum, plasma and other biological fluids.
Specifications
Specifications
0.156 to 10 ng/mL
ELISA Kit (Colorimetric)
This kit recognizes Human TNR in samples. No significant cross-reactivity or interference between Human TNR and analogues was observed.
Human
1 Kit
7143
0.094 ng/mL
Storage is content dependent.
TNR
Biological Fluids, Plasma, Serum