Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human TEX15 Partial ORF (NP_112561.1, 2690 a.a. - 2789 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | NP_112561.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 56154 |
Molecular Weight (g/mol) | 36.74kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-964-426
|
Abnova™
H00056154Q01S |
10 ug |
Each for $499.00
|
|
|||||
89-964-427
|
Abnova™
H00056154Q01L |
25 ug |
Each for $769.00
|
|
|||||
Description
This gene is similar to a mouse gene that is expressed in the testis.
Sequence: PSGLLTTVASTAQGTHSNLLYSQYFTYFAGEPQANGFVPVNGYFQSQIPASNFRQPIFSQYASHQPLPQATYPYLPNRFVPPEVPWVYAPWHQESFHPGHSpecifications
NP_112561.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp434M2415 | |
TEX15 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
56154 | |
TEX15 (Human) Recombinant Protein (Q01) | |
PSGLLTTVASTAQGTHSNLLYSQYFTYFAGEPQANGFVPVNGYFQSQIPASNFRQPIFSQYASHQPLPQATYPYLPNRFVPPEVPWVYAPWHQESFHPGH | |
RUO | |
TEX15 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title