Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TFF2 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $468.50
Specifications
Antigen | TFF2 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 2 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
TFF2 Polyclonal specifically detects TFF2 in Human samples. It is validated for Western Blot.Specifications
TFF2 | |
Western Blot 2 ug/mL | |
Polyclonal | |
Goat | |
Human | |
SML1, SML1trefoil factor 2, SML1, human spasmolytic polypeptide (SP)10spasmolytic protein 1, SP, Spasmolysin, Spasmolytic polypeptide, trefoil factor 2 | |
TFF2 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q03403 | |
7032 | |
E. coli-derived recombinant human TFF2 Glu24-Tyr129 Accession # Q03403 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human TFF2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human TFF3 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title