Learn More
Novus Biologicals™ Human TGF-beta 2 ELISA Kit
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP191255
Catalog No. NBP191255
Add to cart
Description
Description
The Human TGF-beta 2 ELISA Kit (Colorimetric) measures human TGF-beta 2 in cell culture supernatant, serum.
Specifications
Specifications
31.0 - 1,000pg/mL
7042
ELISA Kit
6.6pg/mL
Human (Hu)
TGF-beta 2
BSC-1 cell growth inhibitor, Cetermin, Glioblastoma-derived T-cell suppressor factor, G-TSF, MGC116892, Polyergin, TGF-beta2, TGF-beta-2, transforming growth factor beta-2, transforming growth factor, beta 2
ELISA
Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human TGF-beta2, Biotin-Conjugate anti-human TGF-beta2 monoclonal antibody, Streptavidin-HRP, Human TGF-beta2 Standard, 1ng/mL, 1N HCl (pretreatment of samples), 1N NaOH (pretreatment of samples), Assay Buffer Concentrate 10x, Wash Buffer Concentrate 40x, Substrate Solution (tetramethyl-benzidine), Stop Solution (0.5M H2SO4), Adhesive Films
96 Tests
Cell culture supernatant, serum
Storage of components varies. See protocol for specific instructions.
TGFB2