Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human THOC2 Partial ORF (NP_065182.1, 1 a.a. - 99 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00057187Q01L

 View more versions of this product

Catalog No. 89-964-668


Add to cart

Description

Description

THO2 is part of the TREX (transcription/export) complex, which includes TEX1 (MIM 606929), HPR1 (MIM 606930), ALY (MIM 604171), and UAP56 (MIM 606390).[supplied by OMIM]

Sequence: MFVSDTVLKERLDPETLESLGLIKQSQQFNQKSVKIKTKLFYKQQKFNLLREENEGYAKLIAELGQDLSGSITSDLILENIKSLIGCFNLDPNRVLDVI
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MFVSDTVLKERLDPETLESLGLIKQSQQFNQKSVKIKTKLFYKQQKFNLLREENEGYAKLIAELGQDLSGSITSDLILENIKSLIGCFNLDPNRVLDVI
RUO
THOC2
Wheat Germ (in vitro)
GST
NP_065182.1
Liquid
57187
THOC2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CXorf3/THO2/dJ506G2.1
THOC2
Yes
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit