Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gibco™ Human TIGAR-TAT Recombinant Protein, PeproTech®
E. coli Recombinant Protein
$89.50 - $3325.00
Specifications
Accession Number | Q9NQ88 |
---|---|
For Use With (Application) | Functional Assay |
Formulation | protein with no preservative |
Gene ID (Entrez) | 57103 |
Molecular Weight (g/mol) | 31.6 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
15014T100UG
|
Gibco™
15014T100UG |
100 μg |
Each for $666.00
|
|
|||||
150-14T-1MG
|
Gibco™
15014T1MG |
1 mg |
Each for $3,325.00
|
|
|||||
15014T250UG
|
Gibco™
15014T250UG |
250 μg |
Each for $1,164.00
|
|
|||||
15014T25UG
|
Gibco™
15014T25UG |
25 μg |
Each for $217.00
|
|
|||||
15014T500UG
|
Gibco™
15014T500UG |
500 μg |
Each for $1,996.00
|
|
|||||
150-14T-5UG
|
Gibco™
15014T5UG |
5 μg |
Each for $89.50
|
|
|||||
Description
TIGAR is a p53-inducible enzyme that catalyzes the hydrolysis of fructose-2-6 bisphosphate (F-2-6-BP) to fructose-6-phosphate and inorganic phosphate. F-2-6-BP is a powerful activator of 6-phosphofructose-1 kinase, the rate limiting enzyme of glycolysis. By lowering the intracellular level of F-2-6-BP, TIGAR expression leads to increased glucose processing via the pentose phosphate pathway, the major cellular source for NADPH. Protein transduction using TAT fusion proteins represents an alternative methodology for introducing transcription factors and other intracellular proteins into primary, as well as transformed, cells.Specifications
Q9NQ88 | |
protein with no preservative | |
31.6 | |
E. coli | |
RUO | |
TIGAR | |
Human | |
Recombinant | |
Lyophilized |
Functional Assay | |
57103 | |
Human TIGAR-TAT | |
-20°C | |
<1 EU/μg | |
TIGAR | |
Unconjugated | |
ARFALTVVRH GETRFNKEKI IQGQGVDEPL SETGFKQAAA AGIFLNNVKF THAFSSDLMR TKQTMHGILE RSKFCKDMTV KYDSRLRERK YGVVEGKALS ELRAMAKAAR EECPVFTPPG GETLDQVKMR GIDFFEFLCQ LILKEADQKE QFSQGSPSNC LETSLAEIFP LGKNHSSKVN SDSGIPGLAA SVLVVSHGAY MRSLFDYFLT DLKCSLPATL SRSELMSVTP NTGMSLFIIN FEEGREVKPT VQCICMNLQD HLNGLTETRG GGYGRKKRRQ RRR | |
≥ 95% by SDS-PAGE gel and HPLC analyses. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title