Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human TIM-4 His-tag Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Supplier: R&D Systems™ 9407TM050
This item is not returnable.
View return policy
Description
The Recombinant Human TIM-4 His-tag Protein is derived from HEK293. The Recombinant Human TIM-4 His-tag Protein has been validated for the following applications: Bioactivity.Order Info
Ships at ambient temperature.
Specifications
Q96H15 | |
Lyophilized from a 0.2μm filtered solution in PBS. | |
32 kDa | |
Human embryonic kidney cell,HEK293-derived human TIM-4 protein Glu25-Gln314,with a C-terminal 6-His tag | |
RUO | |
FLJ27515, SMUCKLER | |
Recombinant protein | |
Human | |
Reconstitute at 500μg/mL in sterilePBS. |
Bioactivity | |
91937 | |
50μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
<0.10 EU per 1μg of the protein by the LAL method. | |
0.06 to 0.36μg/mL | |
Unconjugated | |
Recombinant | |
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?
Provide Content Correction