Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TIMM8B Partial ORF (NP_036591, 1 a.a. - 83 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00026521Q01L

 View more versions of this product

Catalog No. 89-964-774


Add to cart

Description

Description

This gene encodes a member of a well-conserved family of proteins with similarity to yeast Tim mitochondrial import proteins. This gene is encoded by a nuclear gene and is transported into the intermembrane space of the mitochondrion. When formed into complexes, these proteins guide membrane-spanning proteins across the mitochondrial intermembrane space before they are added into the mitochondrial inner membrane. This gene is adjacent to succinate dehydrogenase, subunit D (SDHD), in which mutations have been found in affected members of families with hereditary paraganglioma

Sequence: MAELGEADEAELQRLVAAEQQKAQFTAQVHHFMELCWDKCVEKPGNRLDSRTENCLSSCVDRFIDTTLAITSRFAQIVQKGGQ
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
34.87kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MAELGEADEAELQRLVAAEQQKAQFTAQVHHFMELCWDKCVEKPGNRLDSRTENCLSSCVDRFIDTTLAITSRFAQIVQKGGQ
RUO
TIMM8B
Wheat Germ (in vitro)
GST
NP_036591
Liquid
26521
TIMM8B (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DDP2/FLJ21744/MGC102866/MGC117373/TIM8B
TIMM8B
Yes
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit