Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human Tlr8 Partial Orf (Np_619542.1, 723 A.A.-825 A.A.) Recombinant Protein With Gst Tag At N-Terminal.

Recombinant proteins expressed from in vitro wheat germ system. Such proteins mirror the conformation and folding of the native proteins in the eukaryotic biological system.

$360.00 - $548.50

Specifications

Format Liquid
Regulatory Status RUO
Formulation 50 mM Tris-HCI, 10 mM reduced Glutathione, pH=8.0 in the elution buffer.
Accession Number NP_619542.1
Purification Method Glutathione Sepharose 4 Fast Flow
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-032-221 View Documents Abnova™
H00051311Q01S
10 ug Each for $360.00
Add to cart
 
89-032-222 View Documents Abnova™
H00051311Q01L
25 ug Each for $548.50
Add to cart
 
Description

Description

  • Good Folding
  • Excellent amino acids labeling

Immunogen Sequence: RISHLPSGFLSEVSSLKHLDLSSNLLKTINKSALETKTTTKLSMLELHGNPFECTCDIGDFRRWMDEHLNVKIPRLVDVICASPGDQRGKSIVSLELTTCVSD

Specifications

Specifications

Liquid
50 mM Tris-HCI, 10 mM reduced Glutathione, pH=8.0 in the elution buffer.
Glutathione Sepharose 4 Fast Flow
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
RUO
NP_619542.1
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit