Learn More
Abnova™ Human TMC8 Partial ORF (NP_689681.2, 616 a.a. - 674 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | NP_689681.2 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 147138 |
Molecular Weight (g/mol) | 32.23kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-943-697
|
Abnova™
H00147138Q01S |
10 ug |
Each for $499.00
|
|
|||||
89-943-698
|
Abnova™
H00147138Q01L |
25 ug |
Each for $769.00
|
|
|||||
Description
Epidermodysplasia verruciformis (EV) is an autosomal recessive dermatosis characterized by abnormal susceptibility to human papillomaviruses (HPVs) and a high rate of progression to squamous cell carcinoma on sun-exposed skin. EV is caused by mutations in either of two adjacent genes located on chromosome 17q25.3. Both of these genes encode integral membrane proteins that localize to the endoplasmic reticulum and are predicted to form transmembrane channels. This gene encodes a transmembrane channel-like protein with 8 predicted transmembrane domains and 3 leucine zipper motifs. [provided by RefSeq]
Sequence: SQTQANARAIHRLRKQLVWQVQEKWHLVEDLSRLLPEPGPSDSPGPKYPASQASRPQSFSpecifications
NP_689681.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.23kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
EV2/EVER2/EVIN2/FLJ40668/FLJ43684/MGC102701/MGC40121 | |
TMC8 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
147138 | |
TMC8 (Human) Recombinant Protein (Q01) | |
SQTQANARAIHRLRKQLVWQVQEKWHLVEDLSRLLPEPGPSDSPGPKYPASQASRPQSF | |
RUO | |
TMC8 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.