Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TMPRSS11D Full-length ORF (NP_004253.1, 1 a.a. - 418 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$460.50 - $709.50

Specifications

Name TMPRSS11D (Human) Recombinant Protein (P01)
Recombinant Yes
Purification Method Glutathione Sepharose 4 Fast Flow
For Use With (Application) Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Molecular Weight 72.7kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-965-100
View Documents
Abnova™
H00009407P01L
25 ug
Each for $709.50
Add to cart
 
89-965-099
View Documents
Abnova™
H00009407P01S
10 ug
Each for $460.50
Add to cart
 
Description

Description

This gene encodes a trypsin-like serine protease released from the submucosal serous glands onto mucous membrane. It is a type II integral membrane protein and has 29-38% identity in the sequence of the catalytic region with human hepsin, enteropeptidase, acrosin, and mast cell tryptase. The noncatalytic region has little similarity to other known proteins. This protein may play some biological role in the host defense system on the mucous membrane independently of or in cooperation with other substances in airway mucous or bronchial secretions. [provided by RefSeq]

Sequence: MYRPARVTSTSRFLNPYVVCFIVVAGVVILAVTIALLVYFLAFDQKSYFYRSSFQLLNVEYNSQLNSPATQEYRTLSGRIESLITKTFKESNLRNQFIRAHVAKLRQDGSGVRADVVMKFQFTRNNNGASMKSRIESVLRQMLNNSGNLEINPSTEITSLTDQAAANWLINECGAGPDLITLSEQRILGGTEAEEGSWPWQVSLRLNNAHHCGGSLINNMWILTAAHCFRSNSNPRDWIATSGISTTFPKLRMRVRNILIHNNYKSATHENDIALVRLENSVTFTKDIHSVCLPAATQNIPPGSTAYVTGWGAQEYAGHTVPELRQGQVRIISNDVCNAPHSYNGAILSGMLCAGVPQGGVDACQGDSGGPLVQEDSRRLWFIVGIVSWGDQCGLPDKPGVYTRVTAYLDWIRQQTGI
Specifications

Specifications

TMPRSS11D (Human) Recombinant Protein (P01)
Glutathione Sepharose 4 Fast Flow
72.7kDa
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit