Learn More
Novus Biologicals™ Human TNF-alpha ELISA Kit
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP183741
Catalog No. NBP183741
Add to cart
Description
Description
The Human TNF-alpha ELISA Kit (Colorimetric) measures human TNF-alpha in cell culture supernatant, serum, plasma (edta, citrate, heparin).
Specifications
Specifications
23 - 1,500pg/mL
7124
ELISA Kit
Storage of components varies. See protocol for specific instructions.
Adaptive Immunity, Asthma, Autophagy, Biologically Active Proteins, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity
APC1 protein, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necr
Human
ELISA
Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human TNF-alpha, Biotin-Conjugate anti-human TNF-alpha polyclonal antibody, Streptavidin-HRP, Human TNF-alpha Standard lyophilized, 3000 pg/ml upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
5.0pg/mL
TNF-alpha
TNF
96 Tests
Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum