Learn More
R&D Systems™ Human TNF-alpha Quantikine ELISA Kit
Human TNF-alpha Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Manufacturer:  R&D Systems™ PDTA00D
 View more versions of this product
Catalog No. PDTA00D
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
The Human TNF-alpha Quantikine ELISA Kit quantifies human TNF-alpha in cell culture supernates (50 ul), serum (50 ul), plasma (50 ul).
1 Pack (50 Plates)
Specifications
Specifications
1 Pack (50 Plates)
15.60 - 1000 pg/mL
7124
Cell Culture Supernates (50 uL),Serum (50 uL),EDTA Plasma (50 uL),Heparin Plasma (50 uL)
4.0 hours
Human
Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum
ELISA
6.23 pg/mL
Store the unopened product at 2° to 8°C. Do not use past expiration date.
TNF-alpha
APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha