Learn More
GE Healthcare Human TNFα Biotrak Easy ELISA
Enzyme-linked immunosorbent assay for quantitative detection of human TNFα in cell culture supernatants, human serum, plasma, urine, synovial fluid, amniotic fluid, or other body fluids.
Manufacturer:  GE Healthcare RPN5967
Catalog No. 45-005-922
Add to cart
Includes: 1 x 96-well microplate coated with Mab (murine) to (h)TNFα, biotin-conjugate (anti-TNFα), streptavidin-HRP, and sample diluent, lyophilized; 2 x TNFα standard curve; 1 bottle (25mL) 20X wash buffer concentrate (PBS with 1% Tween 20); 15m
Description
Description
  • Based upon format in which all main assay components are supplied lyophilized and preloaded on 96-well microplate
  • Easy ELISAs are easier to use than conventional ELISAs
  • Fewer steps and reduced pipetting results in greater precision
    Order Info
    Each Easy ELISA microplate contains the following components: coating antibody, standard protein, detection antibody, streptavidin-HRP (if necessary), and sample diluent. Only standard wells contain standard protein. Sample wells do not contain standard p
Specifications
Specifications
7.8 to 500 pg/mL
15 min.
Human TNFα Biotrak Easy ELISA
1.65 pg/mL
-20°C
Quantitative detection of human TNFα in cell culture supernatants, human serum, plasma, urine, synovial fluid, amniotic fluid, or other body fluids.
1 x 96-well microplate coated with Mab (murine) to (h)TNFα, biotin-conjugate (anti-TNFα), streptavidin-HRP, and sample diluent, lyophilized; 2 x TNFα standard curve; 1 bottle (25mL) 20X wash buffer concentrate (PBS with 1% Tween 20); 15m
Intra-Assay Precision: 6% CV; Inter-Assay Precision: 9.3% CV
50 μL
210 min.