Learn More
Novus Biologicals™ Human TNNC2 ELISA Kit (Colorimetric)
Human TNNC2 ELISA Kit (Colorimetric) from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Supplier: Novus Biologicals™ NBP276597
Description
The Human TNNC2 ELISA Kit (Colorimetric) measures human TNNC2 in serum, plasma and other biological fluids.Specifications
31.25 to 2000 pg/mL | |
ELISA Kit (Colorimetric) | |
This kit recognizes Human TNNC2 in samples. No significant cross-reactivity or interference between Human TNNC2 and analogues was observed. | |
TNNC2 | |
1 Kit |
18.75 pg/mL | |
Biological Fluids, Plasma, Serum | |
7125 | |
Human | |
Storage of components varies. See protocol for specific instructions. |