Learn More
Crystal Chem IncSupplier Diversity Partner Human Total Bile Acids Assay Kit, 96-tests DFS Item
Manufacturer:  Crystal Chem IncSupplier Diversity Partner 80269
The Total Bile Acids Assay is used to quantify levels of total bile acids in human serum and plasma in less than 10 min.Highlights: Kit uses sample size of only 20 µL. Excellent correlation to ELISA method. Fast, 10 minute procedure. Protocol for liver and feces samples available.Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7684