Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human Total FGF R2 alpha DuoSet IC
contains the basic components required for the development of sandwich ELISAs to measure in cell lysates
Specifications
Product Type | ELISA Development Kit for Intracellular Proteins |
---|---|
Quantity | 1 Kit |
Species | Human |
Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules |
Gene ID (Entrez) | 2263 |
Catalog Number | Mfr. No. | Packaging Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Packaging Quantity | Price | Quantity & Availability | |||||
DYC6655
|
R&D Systems™
DYC6655 |
5 Plates | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
DYC665E
|
R&D Systems™
DYC665E |
15 Plates | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
DYC6652
|
R&D Systems™
DYC6652 |
2 Plates | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
- Optimized capture and detection antibody pairings with recommended concentrations save lengthy development time
- Development protocols are provided to guide further assay optimization
- Assay can be customized to your specific needs
Specifications
ELISA Development Kit for Intracellular Proteins | |
Human | |
2263 |
1 Kit | |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title