Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human Total FGFR3 DuoSet IC ELISA
contains the basic components required for the development of sandwich ELISAs to measure in cell lysates
$545.20 - $997.60
Specifications
Product Type | ELISA Development Kit for Intracellular Proteins |
---|---|
Quantity | 1 Kit |
Species | Human |
Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules |
Gene ID (Entrez) | 2261 |
Catalog Number | Mfr. No. | Packaging Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Packaging Quantity | Price | Quantity & Availability | |||||
DYC7662
|
R&D Systems™
DYC7662 |
2 Plates |
Each for $545.20
|
|
|||||
DYC7665
|
R&D Systems™
DYC7665 |
5 Plates |
Each for $997.60
|
|
|||||
DYC766E
|
R&D Systems™
DYC766E |
15 Plates |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | |||||
Description
Complete ELISpot kits are highly sensitive, microplate-based assays for the detection of cytokine secreting cells. Kits are available for detection and enumeration of a single analyte or two analytes simultaneously. Complete ELISpot kits are ready-to-r
The Human Total FGF R3 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human FGFR3.Specifications
ELISA Development Kit for Intracellular Proteins | |
Human | |
2261 |
1 Kit | |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title