Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human Total Survivin Surveyor IC
Supplier: R&D Systems™ SUV647
This item is not returnable.
View return policy
Description
Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Lysates (100μL)
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Specifications
332 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction