Learn More
Novus Biologicals™ Human TRAIL/TNFSF10 ELISA Kit
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP182418
Catalog No. NBP182418
Add to cart
Description
Description
The Human TRAIL/TNFSF10 ELISA Kit (Colorimetric) measures human TRAIL/TNFSF10 in cell culture supernatant, serum, plasma (edta, citrate, heparin).
Specifications
Specifications
15.6 - 1000pg/mL
8743
ELISA Kit
Storage of components varies. See protocol for specific instructions.
Apoptosis, Cancer, Cell Biology, Signal Transduction, Tumor Suppressors
Apo-2 ligand, APO2L, Apo-2Ltumor necrosis factor (ligand) family, member 10, APO2Ltumor necrosis factor apoptosis-inducing ligand splice variant delta, CD253, CD253 antigen, Protein TRAIL, TL2, TNF-related apoptosis-inducing ligand, TRAILTNF-related apopt
Human
ELISA
Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human TRAIL, Biotin-Conjugate anti-human TRAIL monoclonal antibody, Streptavidin-HRP, Human TRAIL Standard lyophilized, 2ng/mL upon reconstitution, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
5.0pg/mL
TRAIL/TNFSF10
TNFSF10
96 Tests
Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum