Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TRIM22 Full-length ORF (NP_006065.2, 1 a.a. - 498 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Specifications

Name TRIM22 (Human) Recombinant Protein (P01)
Recombinant Yes
Purification Method Glutathione Sepharose 4 Fast Flow
For Use With (Application) Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Molecular Weight 83.3kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-965-659
View Documents
Abnova™
H00010346P01L
25 ug This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
89-965-658
View Documents
Abnova™
H00010346P01S
10 ug This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This protein localizes to the cytoplasm and its expression is induced by interferon. The protein down-regulates transcription from the HIV-1 LTR promoter region, suggesting that function of this protein may be to mediate interferon's antiviral effects. [provided by RefSeq]

Sequence: MDFSVKVDIEKEVTCPICLELLTEPLSLDCGHSFCQACITAKIKESVIISRGESSCPVCQTRFQPGNLRPNRHLANIVERVKEVKMSPQEGQKRDVCEHHGKKLQIFCKEDGKVICWVCELSQEHQGHQTFRINEVVKECQEKLQVALQRLIKEDQEAEKLEDDIRQERTAWKNYIQIERQKILKGFNEMRVILDNEEQRELQKLEEGEVNVLDNLAAATDQLVQQRQDASTLISDLQRRLRGSSVEMLQDVIDVMKRSESWTLKKPKSVSKKLKSVFRVPDLSGMLQVLKELTDVQYYWVDVMLNPGSATSNVAISVDQRQVKTVRTCTFKNSNPCDFSAFGVFGCQYFSSGKYYWEVDVSGKIAWILGVHSKISSLNKRKSSGFAFDPSVNYSKVYSRYRPQYGYWVIGLQNTCEYNAFEDSSSSDPKVLTLFMAVPPCRIGVFLDYEAGIVSFFNVTNHGALIYKFSGCRFSRPAYPYFNPWNCLVPMTVCPPSS
Specifications

Specifications

TRIM22 (Human) Recombinant Protein (P01)
Glutathione Sepharose 4 Fast Flow
83.3kDa
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit