Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TRIM34 Full-length ORF (NP_569073.1, 1 a.a. - 243 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00053840P01L

 View more versions of this product

Catalog No. 89-965-697


Add to cart

Description

Description

The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. Expression of this gene is up-regulated by interferon. This gene is mappped to chromosome 11p15, where it resides within a TRIM gene cluster. Alternate splicing of this gene generates four transcript variants. Additionally, a read-through transcript transcribed from this gene and TRIM6 has been observed. [provided by RefSeq]

Sequence: MELLQDMSGIMKWSEIWRLKKPKMVSKKLKTVFHAPDLSRMLQMFRELTAVRCYWVDVTLNSVNLNLNLVLSEDQRQVISVPIWPFQCYNYGVLGSQYFSSGKHYWEVDVSKKTAWILGVYCRTYSRHMKYVVRRCANRQNLYTKYRPLFGYWVIGLQNKCKYGVFEESLSSDPEVLTLSMAVPPCRVGVFLDYEAGIVSFFNVTSHGSLIYKFSKCCFSQPVYPYFNPWNCPAPMTLCPPSS
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
54.7kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
IFP1/RNF21
TRIM34
Yes
NP_569073.1
Liquid
53840
TRIM34 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MELLQDMSGIMKWSEIWRLKKPKMVSKKLKTVFHAPDLSRMLQMFRELTAVRCYWVDVTLNSVNLNLNLVLSEDQRQVISVPIWPFQCYNYGVLGSQYFSSGKHYWEVDVSKKTAWILGVYCRTYSRHMKYVVRRCANRQNLYTKYRPLFGYWVIGLQNKCKYGVFEESLSSDPEVLTLSMAVPPCRVGVFLDYEAGIVSFFNVTSHGSLIYKFSKCCFSQPVYPYFNPWNCPAPMTLCPPSS
RUO
TRIM34
Wheat Germ (in vitro)
GST
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit