Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TRIM36 Partial ORF (NP_061170, 391 a.a. - 499 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00055521Q01L

 View more versions of this product

Catalog No. 89-965-703


Add to cart

Description

Description

The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. Multiple alternatively spliced transcript variants that encode different protein isoforms have been described for this gene. [provided by RefSeq]

Sequence: TSFEDYVVNTSKQTELLGELSFFSSGIDVPEINEEQSKVYNNALINWHHPEKDKADSYVLEYRKINRDDEMSWNEIEVCGTSKIIQDLENSSTYAFRVRAYKGSICSPC
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.73kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
TSFEDYVVNTSKQTELLGELSFFSSGIDVPEINEEQSKVYNNALINWHHPEKDKADSYVLEYRKINRDDEMSWNEIEVCGTSKIIQDLENSSTYAFRVRAYKGSICSPC
RUO
TRIM36
Wheat Germ (in vitro)
GST
NP_061170
Liquid
55521
TRIM36 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HAPRIN/RBCC728/RNF98
TRIM36
Yes
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit